Learn More
Medchemexpress LLC (2R,3S)-brassinazole | 259200-30-1 | 99.8% | 1 ML

Supplier: Medchemexpress LLC HY121161C10MM 1ML
(2R,3S)-brassinazole is the enantiomeric form of brassinazole, a triazole-type inhibitor of brassinosteroid biosynthesis. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), it is intended for use in plant biology research to modulate growth and study BR signalling pathways.
- Ready-to-use 10 mM solution in DMSO
- High purity 99.8% with enantiomeric excess reported as 100%
- Molecular weight 327.81 g/mol
- Soluble in DMSO for easy preparation of working concentrations
- Suitable for studies of plant growth regulation and brassinosteroid biosynthesis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.