Learn More
Medchemexpress LLC 2R 3S-E1R 100mg | 1424832-60-9 | 10 MG

Supplier: Medchemexpress LLC HY116463A100MG
(2R,3S)-E1R is the (2R,3S) enantiomer of E1R and functions as a sigma-1 receptor positive allosteric modulator (Sig1R PAM) for research into cognition and memory disorders. The compound is provided as a light yellow to yellow solid and is intended for research use only.
- Sigma-1 receptor positive allosteric modulator
- Used in research on cognition and memory disorders
- Molecular formula C13H16N2O2
- Molecular weight 232.28 g/mol
- High purity confirmed by LCMS
- Supplied as a light yellow to yellow solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.