Learn More
Medchemexpress LLC 2R 3S-E1R 10mM 1mL | 1424832-60-9 | 232.28 g·mol⁻1 | C13H16N2O2 | 1 ML

Supplier: Medchemexpress LLC HY116463A10MM 1ML
(2R,3S)-E1R is a sigma-1 receptor positive allosteric modulator used in preclinical research into cognition and memory disorders. It is an enantiomer of E1R supplied as a research-grade chemical for in vitro studies and stock solution preparation.
- Sigma-1 receptor positive allosteric modulator for cognition and memory research.
- Enantiomer of E1R.
- Purity 98.84% (batch example).
- Molecular formula C13H16N2O2.
- Molecular weight 232.28 g·mol⁻1.
- Appearance light yellow to yellow solid.
- High solubility in DMSO (250 mg/mL), suitable for stock solutions.
- Recommended storage: powder at 4°C or -20°C; in solvent at -20°C or -80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.