Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2R 3S-E1R 50mg | 1424832-60-9 | 232.28 g/mol | C13H16N2O2 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY116463A50MG

Encompass_Preferred

(2R,3S)-E1R is the (2R,3S) enantiomer of E1R and functions as a sigma-1 receptor positive allosteric modulator (Sig1R PAM) for preclinical research into cognition and memory disorders. Supplied as a powder for laboratory use (CAS 1424832-60-9; C13H16N2O2; MW 232.28 g/mol). For research use only.

  • Solid powder, light yellow.
  • CAS number 1424832-60-9.
  • Molecular formula C13H16N2O2; molecular weight 232.28 g/mol.
  • Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Reported purity ~98.8% (supplier specification).
  • Intended for preclinical research; not for human or clinical use.

Catalog No. 50-002-27367


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.