Learn More
Medchemexpress LLC HY-116463A 5mg Medchemexpress, (2R,3S)-E1R CAS:1424832-60-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY116463A5MG
Encompass_Preferred
Medchemexpress, HY-116463A 5mg (2R,3S)-E1R CAS:1424832-60-9 (2R,3S)-E1R (Compound 2c) is an enantiomer of E1R. (2R,3S)-E1R is a sigma-1 receptor positive allosteric modulator (Sig1R PAM) for the treatment of cognition/memory disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9421