Learn More
Medchemexpress LLC (2R,3S)-oxirane-2,3-dicarboxylic acid | 16533-72-5 | MFCD00191645 | 98.0% | 132.07 g/mol | C4H4O5 | 500MG
SDP
Supplier:  Medchemexpress LLC HY125791500MG
Encompass_Preferred
cis-Epoxysuccinic acid is a small-molecule succinate receptor (SUCNR1/GPR91) agonist used in biochemical and pharmacological research. It inhibits cAMP levels with an EC50 of 2.7 μM and is supplied as a white to off-white solid for in vitro studies.
  • Acts as SUCNR1 (GPR91) agonist with EC50 = 2.7 μM.
  • High purity: 98.0%.
  • Chemical formula C4H4O5; molecular weight 132.07 g/mol.
  • Available as solid in multiple sizes including 500 mg and as 10 mM solution in DMSO.
  • White to off-white solid suitable for biochemical assays.
Catalog No. 50-002-36406