Learn More
Medchemexpress LLC 2R/S-6-PNG 50mg | 68682-01-9 | 50 MG

Supplier: Medchemexpress LLC HY11568150MG
2R/S-6-prenylnaringenin is a prenylated flavonoid research chemical that acts as a reversible Cav3.2 T-type calcium channel blocker and is used for in vitro and in vivo neuroscience studies. For research use only; not for human or clinical use.
- Acts as a Cav3.2 T-type calcium channel blocker.
- High purity: 99.88%.
- Chemical formula C20H20O5; molecular weight 340.37 g/mol.
- Soluble in DMSO (125 mg/mL); ultrasonic recommended.
- Available in multiple pack sizes, including 50 MG.
- Suggested storage: powder -20°C (up to 3 years); in solvent -80°C (6 months), -20°C (1 month).
- Intended for research use only; not for human or clinical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.