Learn More
Medchemexpress LLC (2RS)-2-(4-Formylphenyl)propionic acid | 43153-07-7 | MFCD08275588 | >98.0% | 178.18 g·mol⁻¹ | C10H10O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1312605MG
Encompass_Preferred
Ibuprofen Impurity K is an analytical standard of 2-(4-formylphenyl)propionic acid (CAS 43153-07-7) used for impurity profiling, method development, and analytical testing of ibuprofen formulations. It is supplied in laboratory-scale quantities for research and quality-control applications; storage and handling instructions are provided in the certificate of analysis.
  • Analytical standard for impurity profiling and method validation.
  • Relevant to COX pathway studies and ibuprofen degradation analysis.
  • Available in small, laboratory-friendly quantities (e.g., 5 mg).
  • Reported high purity for reliable analytical results.
  • Includes storage and shipping guidance; consult the certificate of analysis.
  • Suitable for research and quality-control applications.
Catalog No. 50-002-62933