Learn More
Medchemexpress LLC (2S)-1-(6-chloro-5-fluoroindol-1-yl)propan-2-amine | 169675-08-5 | 99.5% | 226.68 g/mol | C11H12ClFN2 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12383850MG
Encompass_Preferred
Ro60-0175 is a selective serotonin 5-HT2C receptor agonist used as a research tool in preclinical studies to probe serotonergic signaling and behavioral outcomes such as reduced cocaine self-administration. The compound is supplied as a solid research chemical and is used in vitro and in vivo with appropriate formulation and storage precautions.
Catalog No. 50-002-37629