Learn More
Medchemexpress LLC (2S)-2-(6-Methoxy-2-naphthyl)propanoic acid sodium salt | 26159-34-2 | C14H13NaO3 | 10 G

Supplier: Medchemexpress LLC HY15030A10G
Naproxen sodium is a COX-1 and COX-2 inhibitor, demonstrating IC50s of 8.72 and 5.15 μM respectively in cell assays. This product is intended for research use only and has not been fully validated for medical applications.
- Highly pure with 99.74% (LCMS)
- Appears as a white to off-white solid
- Suitable for research in inflammation/immunology, cancer, autophagy, and COX pathways
- Store at 4°C, sealed, away from moisture; in solvent, store at -80°C for 6 months or -20°C for 1 month
- Molecular weight of 252.24
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.