Learn More
Medchemexpress LLC (2S)-2-(6-Methoxy-2-naphthyl)propanoic acid sodium salt | 26159-34-2 | C14H13NaO3 | 25 G

Supplier: Medchemexpress LLC 25GHY15030A
Naproxen sodium is a nonsteroidal anti-inflammatory drug (NSAID) and a potent inhibitor of both COX-1 and COX-2 enzymes. It demonstrates anti-inflammatory and antifibrotic effects, downregulating TGF-β levels and Smad3/4 complex formation, and inhibiting pain, fever, and PGE2 time-courses.
- Purity of 99.74%
- White to off-white solid appearance
- Store at 4°C, sealed, away from moisture
- When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture
- Inhibits COX-1 (IC50 of 9.55 μM in intact cells)
- Inhibits COX-2 (IC50 of 5.65 μM in intact cells)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.