Learn More
Medchemexpress LLC (2S)-2-amino-4-phosphonobutanoic acid | 23052-81-5 | MFCD00083244 | 99.0% | 183.10 g/mol | C4H10NO5P | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100781A10MG
Encompass_Preferred
L-AP4 ((2S)-2-amino-4-phosphonobutanoic acid) is a selective agonist of group III metabotropic glutamate receptors (mGluRs) used in neuroscience research to investigate mGluR-mediated signaling and synaptic modulation. It displays submicromolar potency at mGlu4 and mGlu8 and differing activity across other group III subtypes.
Catalog No. 50-002-13410