Learn More
Medchemexpress LLC (2S)-2-amino-6-[[(1S,8R)-9-bicyclo[6.1.0]non-4-ynyl]methoxycarbonylamino]hexanoic acid | 1384100-45-1 | 95.0% | 322.40 g/mol | C17H26N2O4 | 10mg
SDP
Supplier:  Medchemexpress LLC HYW75458810MG
Encompass_Preferred
exo-BCN-L-Lysine is a Click Amino Acid that can be used as a linker in the synthesis of PROTAC molecules exo-BCN-L-Lysine contains a BCN group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups
Catalog No. 50-002-70346