Learn More
Medchemexpress LLC (2S)-2'-Methoxykurarinone | 270249-38-2 | 452.54 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN174610MM/1ML
Encompass_Preferred
(2S)-2'-Methoxykurarinone is a compound isolated from the roots of Sophora flavescens. It exhibits anti-inflammatory, antipyretic, antidiabetic, and antineoplastic effects. It also inhibits osteoclastogenesis and bone resorption by down-regulating RANKL signaling and shows cytotoxic activity against human myeloid leukemia HL-60 cells.
  • Anti-inflammatory effects
  • Antipyretic effects
  • Antidiabetic effects
  • Antineoplastic effects
  • Inhibits osteoclastogenesis and bone resorption through down-regulation of RANKL signaling
  • Displays cytotoxic activity against human myeloid leukemia HL-60 cells
Catalog No. 50-004-15190