Learn More
TARGETMOL CHEMICALS INC 2S-2-Methoxykurarinone 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN12155MG
Encompass_Preferred
Also available in 1 mg and bulk. Please contact Fisher for quotes. (2S)-2'-Methoxykurarinone is isolated from the roots of Sophora flavescens. (2S)-2'-Methoxykurarinone has anti-inflammatory, antipyretic, antidiabetic, and antineoplastic effects. (2S)-2'-Methoxykurarinone inhibits osteoclastogenesis and bone resorption through down-regulation of RANKL signaling. (2S)-2'-Methoxykurarinone displays cytotoxic activity against human myeloid leukemia HL-60 cells. Purity 98.17%
Catalog No. 50-287-7172