Learn More
Medchemexpress LLC HY-N1746 5mg Medchemexpress, (2S)-2'-Methoxykurarinone CAS:270249-38-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN17465MG
Encompass_Preferred
Medchemexpress, HY-N1746 5mg (2S)-2'-Methoxykurarinone CAS:270249-38-2 (2S)-2'-Methoxykurarinone, a compound isolated from the roots of Sophora flavescens, has anti-inflammatory, antipyretic, antidiabetic, and antineoplastic effects. (2S)-2'-Methoxykurarinone (MK) inhibits osteoclastogenesis and bone resorption through down-regulation of RANKL signaling. (2S)-2'-Methoxykurarinone (MK) displays cytotoxic activity against human myeloid leukemia HL-60 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0349