Learn More
Medchemexpress LLC (2S)-2-pyrrolidinemethanol | 23356-96-9 | MFCD00005255 | 99.4% | 101.15 g/mol | C5H11NO | 25g
SDP
Supplier:  Medchemexpress LLC HYY112625G
Encompass_Preferred
(2S)-2-Pyrrolidinemethanol ((S)-( )-Prolinol) reveals binucleophilic character and can be anchored to phthalonitrile derivative from both nitrogen and oxygen atoms (2S)-2-Pyrrolidinemethanol can be utilized as chiral catalyst[1][2]
Catalog No. 50-002-66791