Learn More
Medchemexpress LLC (2S,3R)-2,3,4-Trihydroxybutanal-13C-2 | 90913-09-0 | 99.4% | 121.10 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW012864S210MG
Encompass_Preferred
(2S,3R)-2,3,4-Trihydroxybutanal-13C-2 is the 13C labeled (2S,3R)-2,3,4-Trihydroxybutanal. This compound can be used as a tracer and as an internal standard for quantitative analysis. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54808