Learn More
Medchemexpress LLC (2S,3R)-2,3,4-Trihydroxybutanal-13C-2 | 90913-09-0 | 99.4% | 121.10 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW012864S2100MG
Encompass_Preferred
(2S,3R)-2,3,4-Trihydroxybutanal-13C-2 is the 13C labeled (2S,3R)-2,3,4-Trihydroxybutanal. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-54849