Learn More
Medchemexpress LLC (2S,3R,4S)-4-Hydroxyisoleucine | 55399-93-4 | 98.0% | 147.17 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01027150MG
Encompass_Preferred
(2S,3R,4S)-4-Hydroxyisoleucine is an orally active compound isolated from *Trigonella foenum-graecum*. It exhibits anti-diabetic and anti-diabetic nephropathy activity, significantly increasing insulin release in isolated rat pancreatic islets and showing non-insulin dependent anti-diabetic activity in type I diabetic rats. It induces a reduction in blood glucose, restores diabetic lipid profiles, reverses HDL changes, and reduces hyperuricemia.
  • Significantly increases insulin release
  • Demonstrates anti-diabetic activity
  • Restores diabetic lipid profile
  • Reduces increased hyperuricemia
  • Isolated from *Trigonella foenum-graecum*
Catalog No. 50-003-66534