Learn More
Medchemexpress LLC (2S,3R)-brassinazole | 259200-31-2 | 98.4% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY121161B10MG
Encompass_Preferred
(2S,3R)-Brassinazole is the enantiomer of brassinazole, a triazole-type inhibitor of brassinosteroid biosynthesis used in plant biology research to modulate plant growth and development. It is supplied as a white to off-white solid with high chemical purity and is typically dissolved in DMSO for in vitro assays.
  • High purity suitable for research applications.
  • Enantiomeric excess of 100% for stereospecific studies.
  • Soluble in DMSO (20 mg/mL) with sonication and warming.
  • Offered in small solid quantities ideal for assay development.
  • Structural identifiers (SMILES) available for cheminformatics use.
Catalog No. 50-002-31741