Learn More
Medchemexpress LLC (2s,3r)-brassinazole | 259200-31-2 | 98.4% | 5 MG

Supplier: Medchemexpress LLC HY121161B5MG
(2S,3R)-Brassinazole is the enantiomer of brassinazole, a triazole-type inhibitor of brassinosteroid biosynthesis used in plant biology research. It interferes with oxidative steps from 6-oxo-campestanol to teasterone and is reported to be the more active enantiomer.
- Enantiomer of brassinazole that inhibits brassinosteroid biosynthesis.
- CAS number 259200-31-2.
- Purity 98.39% (ee. 100%).
- Molecular weight 327.81 g·mol⁻¹; formula C18H18ClN3O.
- Solid, white to off-white; soluble in DMSO (~20 mg/mL).
- Intended for research use in plant growth and development studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.