Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S,3R)-ethyl 2-amino-3-hydroxybutanoate hydrochloride | 39994-70-2 | MFCD07437964 | ≥97.0% | 183.63 | C6H14ClNO3 | 100 G
SDP

Supplier:  Medchemexpress LLC HYW040333100G

Encompass_Preferred

(2S,3R)-ethyl 2-amino-3-hydroxybutanoate hydrochloride is a chiral threonine-derived amino acid ester supplied as the hydrochloride salt for research use. It is a viscous liquid with defined physical and chemical properties suitable for synthetic and analytical applications.

  • Chemical name: (2S,3R)-ethyl 2-amino-3-hydroxybutanoate hydrochloride.
  • CAS number: 39994-70-2.
  • Molecular formula: C6H14ClNO3.
  • Molecular weight: 183.63 g/mol.
  • Purity: ≥97.0%.
  • Appearance: viscous liquid.
  • Storage: 4°C, sealed and away from moisture; in solvent -80°C (6 months) or -20°C (1 month).
  • Available pack sizes: 10 g, 25 g, 100 g, and larger quantities by quote.

Catalog No. 50-002-09376


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.