Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2S,3R)-ethyl 2-amino-3-hydroxybutanoate hydrochloride | 39994-70-2 | MFCD07437964 | ≥97.0% | 183.63 g/mol | C6H14ClNO3 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW040333

Encompass_Preferred

(2S,3R)-Ethyl 2-amino-3-hydroxybutanoate hydrochloride is the hydrochloride salt of the ethyl ester of the amino acid threonine, supplied as a research reagent for synthetic chemistry and biochemical applications. It is provided as a viscous liquid with high enantiomeric definition for use as a chiral building block or reference material.

  • Purity ≥97.0%.
  • Molecular weight 183.63 g/mol.
  • Molecular formula C6H14ClNO3.
  • Appearance viscous liquid; form: liquid.
  • Storage 4°C, sealed and away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).
  • Available in laboratory pack sizes, e.g., 25 G.

Catalog No. 50-002-09668


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.