Learn More
Medchemexpress LLC 2S 3R-LP99 5mg | 1808948-28-8 | 516.05 g/mol | C26H30ClN3O4S | 5 MG

Supplier: Medchemexpress LLC HY1162275MG
(2S,3R)-LP99 is a selective small-molecule inhibitor of BRD7 and BRD9 provided as a white to off-white solid for research use. It is supplied with analytical documentation to support experimental reproducibility.
- High purity (99.4%) suitable for biochemical and cellular assays.
- Molecular weight 516.05 g/mol and formula C26H30ClN3O4S.
- Supplied as a solid with recommended storage at -20°C (powder) and guidance for solvent storage.
- Includes datasheet, certificate of analysis, and SDS for characterization and handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.