Learn More
Medchemexpress LLC (2S,3R)-Voruciclib hydrochloride | 2920469-94-7 | 98.0% | 506.30 g·mol⁻¹ | C22H20Cl2F3NO5 | 50MG

Supplier: Medchemexpress LLC HY12422B50MG
(2S,3R)-Voruciclib hydrochloride is the enantiomer of voruciclib hydrochloride, an orally active cyclin-dependent kinase (CDK) inhibitor used in research to probe CDK-mediated signaling and cell-cycle regulation. The compound is supplied as a light yellow to yellow solid, with molecular formula C22H20Cl2F3NO5, molecular weight 506.30 g·mol⁻¹, and typical purity of 98.0% by NMR. Recommended storage is sealed at 4 °C; solutions may be stored at -80 °C for 6 months or -20 °C for 1 month.
- Enantiomer of voruciclib hydrochloride.
- Orally active cyclin-dependent kinase inhibitor used for research.
- Molecular weight 506.30 g·mol⁻¹.
- Chemical formula C22H20Cl2F3NO5.
- Purity 98.0% by NMR.
- Appearance light yellow to yellow solid.
- Storage: sealed at 4 °C; solutions -80 °C (6 months) or -20 °C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.