Learn More
Medchemexpress LLC (2S,3S)-2-acetamido-3-methylpentanoic acid | 3077-46-1 | MFCD00066072 | 98.8% | 173.21 g/mol | C8H15NO3 | 100 G

Supplier: Medchemexpress LLC HYW016835100G
(2S,3S)-2-Acetamido-3-methylpentanoic acid is an N-acetylated isoleucine derivative supplied as a high-purity amino acid reagent for biochemical and analytical research. It is provided as a white to off-white solid and is suitable for workflows requiring an acetylated isoleucine analog.
- Purity 98.8%.
- Molecular formula C8H15NO3 and molecular weight 173.21 g/mol.
- Appearance white to off-white solid.
- Solubility in DMSO 14 mg/mL; ultrasonic and warming recommended.
- Powder storage: -20°C (3 years) or 4°C (2 years); in solvent short-term storage at -80°C or -20°C.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.