Learn More
Medchemexpress LLC (2S,3S)-2-amino-3-methylpentan-1-ol | 24629-25-2 | MFCD00004731 | 99.9% | 117.19 | C6H15NO | 50 G

Supplier: Medchemexpress LLC HYW01587550G
L-Isoleucinol ((2S,3S)-2-amino-3-methylpentan-1-ol) is a high-purity amino alcohol used as a reagent and chiral building block in chemical synthesis and analytical workflows. It is commonly employed in life-science research for synthesis, derivatization, and method development.
- Amino alcohol useful as a chiral building block.
- Suitable for organic synthesis and derivatization.
- High purity (99.88%).
- Molecular formula C6H15NO; molecular weight 117.19 g/mol.
- CAS 24629-25-2 for unambiguous identification.
- Commonly supplied in 50 g packaging for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.