Learn More
Strem, An Ascensus Company CAS# 2416226-68-9. 500mg. 2-((2S,3S)-3-(tert-Butyl)-4-(3,5-dicyclopentyl-2,6-dimethoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphol-2-yl)propan-2-ol, min. 97%, (99% ee) (2S,3S)-Baryphos.
SDP
Supplier:  Strem, An Ascensus Company 156911500MG
Encompass
CAS# 2416226-68-9. 500mg. 2-((2S,3S)-3-(tert-Butyl)-4-(3,5-dicyclopentyl-2,6-dimethoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphol-2-yl)propan-2-ol, min. 97%, (99% ee) (2S,3S)-Baryphos. . Molecular Weight: 524.69. Molecular Formula: C32H45O4P. Color/form: white solid Strem# 15-6911. http://www.strem.com/catalog/v/15-6911/
Catalog No. 50-206-5430