Learn More
Medchemexpress LLC 2S 3S-E1R 10mM 1mL | 1424832-57-4 | 232.28 g/mol | C13H16N2O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY116463B10MM 1ML
Encompass_Preferred
(2S,3S)-E1R is an enantiomer of E1R that functions as a sigma-1 receptor positive allosteric modulator (Sig1R PAM). It is supplied for research use and is available both as solid quantities and as a ready-to-use solution for in vitro and preclinical studies.
Catalog No. 50-002-26199