Learn More
Medchemexpress LLC (2S)-4-(benzyloxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxobutanoic acid | 86060-84-6 | 445.46 | 25 G

Supplier: Medchemexpress LLC 25GHYW002301
(2S)-4-(benzyloxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxobutanoic acid is an aspartic acid derivative. It is for research use only and not sold to patients. Amino acids and amino acid derivatives, such as this product, have been commercially used as ergogenic supplements due to their influence on anabolic hormone secretion, fuel supply during exercise, mental performance during stress-related tasks, and prevention of exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
- Appearance: Solid
- Color: White to off-white
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
- Soluble in DMSO at 100 mg/mL
- Classified as a peptide, reagent for peptide synthesis, and amino acid derivative
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.