Learn More
Medchemexpress LLC (2S,4R)-1-((S)-17-amino-2-(tert-butyl)-4-oxo-6,9,12,15-tetraoxa-3-azaheptadecanoyl)-4-hydroxy-N-(4-(4-m | 2010159-57-4 | MFCD31657338 | 99.5% | 663.83 g/mol | C32H49N5O8S | 50 MG

Supplier: Medchemexpress LLC HY103604A50MG
(S,R,S)-AHPC-PEG4-NH2 is a von Hippel-Lindau (VHL) E3 ligase ligand-linker conjugate that couples an (S,R,S)-AHPC VHL ligand to a four-unit polyethylene glycol (PEG4) linker terminated in a primary amine. It is supplied as a white to off-white solid and is intended for use as a building block in PROTAC synthesis to enable conjugation of target-binding warheads.
- Provides a VHL-binding ligand with a PEG4 spacer and terminal primary amine for conjugation.
- High purity by LCMS (≈99.5%) suitable for research applications.
- Soluble in DMSO at high concentration (≈170 mg/mL); ultrasonic assistance may be required.
- Molecular weight approximately 663.83 g/mol and formulated as a solid for easy handling.
- Stable when stored as a powder at -20°C (long term) or 4°C (short term); store solutions at -80°C for extended stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.