Learn More
Medchemexpress LLC (2S,4R)-4-hydroxy-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid | 13504-85-3 | MFCD00037329 | 99.9% | 265.26 g/mol | C13H15NO5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0160425G
Encompass_Preferred
N-Cbz-hydroxy-L-proline is a Cbz-protected hydroxy derivative of L-proline used as a chiral building block and research reagent for organic synthesis and life-science studies. Supplied as a white to off-white solid, it includes analytical documentation and handling guidance for storage and synthetic use.
  • High purity (99.9%).
  • White to off-white solid, easy to weigh and handle.
  • Suitable as a chiral building block for peptide and small-molecule synthesis.
  • Soluble in DMSO (≈100 mg/mL) with ultrasonic aid.
  • Provided with analytical documents (COA, HNMR, RP-HPLC, SDS).
  • Stable when stored refrigerated or frozen per handling instructions.
Catalog No. 50-003-01006