Learn More
Medchemexpress LLC (2s,4r)-4-Hydroxypyrrolidine-2-carboxylic acid | 51-35-4 | 99.9% | 131.13 | 5 G
SDP
Supplier:  Medchemexpress LLC HY401355G
Encompass_Preferred
L-Hydroxyproline is a useful chiral building block employed in the production of various pharmaceuticals. It finds widespread application across medicine, biochemistry, food, cosmetic, and other industrial sectors. It is also a component of certain secondary metabolites, such as actinomycins and echinocandins. Industrially, L-Hydroxyproline is primarily produced via the acid hydrolysis of mammalian collagen, owing to its rich presence in collagen.
  • Chiral building block in pharmaceutical production
  • Used in medicine, biochemistry, food, and cosmetic industries
  • Component of secondary metabolites
  • Manufactured by acid hydrolysis of mammalian collagen
Catalog No. 50-004-05100