Learn More
Medchemexpress LLC (2S,5R)-6-(Benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxamide | 1192651-49-2 | >99.95% | C14H17N3O3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY79858500MG
Encompass_Preferred
This product is (2S,5R)-6-(Benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxamide, an organic compound primarily recognized for its role as an intermediate in the synthesis of Avibactam.
  • Serves as a key intermediate in pharmaceutical synthesis
  • High purity for reliable research and development applications
  • Chemical structure includes a diazabicyclo[3.2.1]octane core
Catalog No. 50-004-33182