Learn More
Medchemexpress LLC (2S,5R)-6-(Benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxamide | 1192651-49-2 | C₁₄H₁₇N₃O₃ | 1 G
SDP
Supplier:  Medchemexpress LLC HY798581G
Encompass_Preferred
(2S,5R)-6-(Benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxamide is a solid chemical designed for laboratory use and the manufacture of substances. With a molecular weight of 275.31, it requires careful handling due to potential hazards. It is important to store this product properly to maintain its stability and efficacy.
  • Harmful if swallowed
  • Causes skin and serious eye irritation
  • May cause respiratory irritation
  • Keep container tightly sealed in cool, well-ventilated area
  • Protect from direct sunlight and ignition sources
  • Recommended powder storage: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • For research use only by qualified personnel
  • Not fully validated for medical applications
Catalog No. 50-004-32750