Learn More
Medchemexpress LLC (2S,5R,6R)-Ticarcillin disodium | 4697-14-7 | 98.2% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB117510MM/1ML
Encompass_Preferred
(2S,5R,6R)-Ticarcillin disodium is a semisynthetic antibiotic that exhibits bactericidal activities. It is identified as the (2S,5R,6R)-enantiomer of Ticarcillin. In vitro, this compound demonstrates reduced activity against Gram-positive cocci, while showing increased activity against the majority of Gram-negative bacilli.
  • It is a semisynthetic antibiotic.
  • It possesses bactericidal activities.
  • It is the (2S,5R,6R)-enantiomer of Ticarcillin.
  • It is less active against Gram-positive cocci.
  • It is more active against most Gram-negative bacilli.
Catalog No. 50-004-11263