Learn More
Medchemexpress LLC (2S,5S)-Methyl 5-phenylpyrrolidine-2-carboxylate hydrochloride | 2170170-16-6 | MFCD22199190 | 99.85% | 241.71 | 1 G

Supplier: Medchemexpress LLC HY233261G
(2S,5S)-Methyl 5-phenylpyrrolidine-2-carboxylate hydrochloride is a drug intermediate for the synthesis of various active compounds.
- Drug intermediate for synthesis of various active compounds.
- Formula: C12H16ClNO2
- Research area: Others
- Target: Drug intermediate
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.