Learn More
Medchemexpress LLC (2S)-6-prenylnaringenin | 68236-13-5 | MFCD21364780 | 99.8% | 340.37 | C20H20O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1071981MG
Encompass_Preferred
(2S)-6-Prenylnaringenin is a prenylated flavonoid research compound that acts as a positive allosteric modulator of GABAA receptors at the α+β- binding interface. It is provided as a high-purity reference material for in vitro pharmacology and neuroscience studies, with molecular formula C20H20O5 and molecular weight 340.37 g/mol.
  • Acts as a GABAA positive allosteric modulator at the α+β- binding interface.
  • High reported purity (99.84%), suitable for biochemical and pharmacological assays.
  • Supplied in small, lab-scale quantities for screening and mechanistic studies.
  • Prenylated flavonoid scaffold useful for structure-activity relationship investigations.
  • Characterized compound with defined molecular formula and molecular weight for analytical work.
Catalog No. 50-002-41468