Learn More
Medchemexpress LLC (2S,6S)-hydroxynorketamine hydrochloride | 1430202-70-2 | MFCD30720898 | 99.3% | 276.16 g/mol | C12H15Cl2NO2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10195525MG
Encompass_Preferred
2S,6S-hydroxynorketamine hydrochloride is the hydrochloride salt of the (2S,6S) stereoisomer of hydroxynorketamine, a ketamine metabolite used in biochemical and pharmacological research. It is supplied as a white to off-white solid and intended for laboratory studies of metabolism and mechanism of action. Handle per appropriate safety protocols.
  • High purity suitable for research applications.
  • Hydrochloride salt form enhances stability and handling.
  • Supplied as a white to off-white solid for easy weighing.
  • Available in small research pack sizes for experimental use.
  • Used to study ketamine metabolism and related pharmacology.
Catalog No. 50-002-48716