Learn More
TARGETMOL CHEMICALS INC 2S-Isoxanthohumol 20MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4S099920MG
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. 1. (2S)-Isoxanthohumol shows an antiviral activity towards herpes viruses (HSV1 and HSV2) and bovine viral diarrhea virus (BVDV). 2. Isoxanthohumol is a polyphenol with antioxidant, anti-inflammatory, and antiangiogenic properties, seems to regulate in vivo vascular proliferation and stabilization and the EC-VSMC-inflammatory crosstalk. Purity 98.82%
Catalog No. 50-287-6679