Learn More
Medchemexpress LLC (2S)-N-(3,5-Dimethylphenyl)-1-[(4-methoxyphenyl)sulfonyl]-2-pyrrolidinecarboxamide | 1361321-96-1 | MFCD28502270 | 99.5% | 388.48 g/mol | C20H24N2O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10183410MG
Encompass_Preferred
ACT-462206 is an orally active, brain-penetrant dual orexin 1/2 receptor antagonist used in neuroscience research to probe orexin signaling and to evaluate behavioral pharmacology. It shows low-nanomolar potency at orexin receptors and is supplied as a high-purity research chemical available in both solid form and as a DMSO solution.
  • Potent dual orexin receptor antagonist with low-nanomolar activity.
  • Brain-penetrant profile suitable for in vivo studies.
  • High purity for reproducible experimental results.
  • Available as solid and DMSO solution for dosing flexibility.
  • Stable when stored under recommended conditions and protected from light.
  • Applicable for pharmacological and behavioral neuroscience research.
Catalog No. 50-002-49301