Learn More
Medchemexpress LLC (2S)-N-(3,5-Dimethylphenyl)-1-[(4-methoxyphenyl)sulfonyl]-2-pyrrolidinecarboxamide | 1361321-96-1 | MFCD28502270 | 99.5% | 388.48 g/mol | C20H24N2O4S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101834100MG
Encompass_Preferred
ACT-462206 is a brain-penetrant dual orexin receptor antagonist used in preclinical research on insomnia, anxiety-related disorders, and addiction. It exhibits potent activity at orexin receptors and is supplied as a high-purity research chemical for laboratory studies. Not for human or veterinary use.
  • Dual orexin receptor antagonist (OX1 and OX2)
  • Brain-penetrant compound suitable for CNS research
  • Potent activity: IC50 ≈ 60 nM (OX1), ≈ 11 nM (OX2)
  • High purity suitable for laboratory use
  • Storage: protect from light; in solvent store at -80°C (6 months) or -20°C (1 month)
  • Available as solid or as 10 mM solution in DMSO
Catalog No. 50-002-48208