Learn More
Medchemexpress LLC (2S)-N-(3,5-dimethylphenyl)-1-[(4-methoxyphenyl)sulfonyl]-2-pyrrolidinecarboxamide | 1361321-96-1 | MFCD28502270 | 99.5% | 388.48 g/mol | C20H24N2O4S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10183450MG
Encompass_Preferred
ACT-462206 is an orally active, brain-penetrant dual orexin 1/orexin 2 receptor antagonist supplied as a high-purity research reagent. It exhibits nanomolar potency (IC50 ≈ 60 nM for OX1 and 11 nM for OX2) and is used in preclinical studies of sleep regulation, anxiety-related disorders, and addiction.
  • Dual orexin 1/2 receptor antagonist with nanomolar potency.
  • Brain-penetrant compound suitable for central nervous system studies.
  • High purity (≈99.5%) for reliable experimental results.
  • Available in small-quantity packs for research use.
  • Stable under recommended storage conditions to preserve integrity.
Catalog No. 50-002-48904