Learn More
Medchemexpress LLC (2S)-N-(3,5-dimethylphenyl)-1-(4-methoxyphenyl)sulfonylpyrrolidine-2-carboxamide | 1361321-96-1 | MFCD28502270 | 99.5% | 388.48 g/mol | C20H24N2O4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10183410MM 1ML
Encompass_Preferred
A potent, orally active dual orexin receptor antagonist for preclinical research. Reported to be brain-penetrant, it is used in studies of sleep, stress/anxiety, and addiction, and is supplied as a concentrated DMSO stock suitable for in vitro and in vivo dosing.
  • Dual orexin 1/orexin 2 receptor antagonist with nanomolar potency.
  • Brain-penetrant profile suitable for CNS studies.
  • Applicable to insomnia, anxiety, and addiction research.
  • Available as a 10 mM stock solution in DMSO for convenient use.
  • High purity (~99.5%) for reproducible experimental results.
Catalog No. 50-002-48128