Learn More
Cayman Chemical 2S-NIcotIn-1-oxIde 10mg
SDP
Supplier:  Cayman Chemical 2215310 MG
Encompass_Preferred
A nicotine metabolite; formed when nicotine is metabolized by CYP2A6 in the liver; has been detected in human semen and sperm
Catalog No. 50-298-6424