Learn More
Medchemexpress LLC (2S)-Octyl-α-hydroxyglutarate | 1391194-64-1 | MFCD09836372 | ≥98.0% | 260.33 | C13H24O5 | 50 MG

Supplier: Medchemexpress LLC HY103641A50MG
(2S)-Octyl-α-hydroxyglutarate is a cell-permeable, octyl-ester modified form of the S-isomer of 2-hydroxyglutarate intended for research use. It serves as a membrane-permeant precursor and analogue of L-2-hydroxyglutarate for biochemical and cell-based studies, supplied as a laboratory reagent.
- Membrane-permeant precursor of S-2-hydroxyglutarate.
- Chemical formula C13H24O5.
- Molecular weight 260.33 g/mol.
- Available as a solid 50 mg pack for research applications.
- Purity reported on the product page (conflicting entries of 95.0% and ≥98.0%).
- For research use only; not for clinical or therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.