Learn More
Medchemexpress LLC (2S)-Octyl-α-hydroxyglutarate | 1391194-64-1 | MFCD29045545 | 95.0% | 260.33 | C13H24O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY103641A1MG
Encompass_Preferred
A cell-permeable octyl-ester derivative of the S-isomer of 2-hydroxyglutarate intended for research use. It is used to deliver intracellular 2-hydroxyglutarate activity in cell-based studies and has been reported to increase DNA double-strand breaks in treated cells. For research use only.
  • Cell-permeable ester to enhance cellular uptake.
  • Models intracellular S-2-hydroxyglutarate activity.
  • Useful for studying DNA damage and metabolic signaling pathways.
  • Available in small research pack sizes for assay development.
Catalog No. 50-002-46899