Learn More
Medchemexpress LLC (2S)-octyl-α-hydroxyglutarate | 1391194-64-1 | MFCD29045545 | ≥98.0% | 260.33 g·mol⁻1 | C13H24O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY103641A25MG
Encompass_Preferred
(2S)-Octyl-α-hydroxyglutarate is the octyl-ester derivative of the S-isomer of 2-hydroxyglutarate. It is used as a membrane-permeable research reagent and precursor to 2-hydroxyglutarate in cellular and biochemical studies, and is supplied as a solid with high reported purity for laboratory research (not for human use).
  • Membrane-permeable precursor of 2-hydroxyglutarate.
  • Supports cell-based and biochemical assays of metabolic pathways.
  • Supplied as a solid with high reported purity (≥98%).
  • Soluble in DMSO for assay preparation.
  • Available in small milligram quantities for research workflows.
Catalog No. 50-002-45098