Learn More
Medchemexpress LLC (2S)-OMPT (triethylamine) | 1217471-69-6 | 98.0% | 1 MG

Supplier: Medchemexpress LLC HY1209671MG
A 98% solution of (2S)-OMPT (triethylamine) in ethanol:chloroform (1:1) used as a chiral ligand for asymmetric catalysis and enantioselective synthesis.
- Chiral ligand for asymmetric catalysis and enantioselective synthesis.
- 98.0% purity.
- Supplied as a solution in ethanol:chloroform (1:1) for immediate use.
- Molecular weight 668.99 g/mol; formula C34H73N2O6PS.
- Store at -20°C; solution stable for up to 2 years.
- Available in a 1 mg vial for small-scale research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.